Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoic Acid Ammonium Salt 99%, Thermo Scientific™

Catalog No. ac401902500
Click to view available options
Quantity:
250 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1863-63-4
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.154
InChI Key VWSRWGFGAAKTQG-UHFFFAOYSA-N
Synonym ammonium benzoate, benzoic acid, ammonium salt, ammonii benzoas, ammonium benzoicum, vulnoc ab, benzoic acid ammonium salt, hsdb 555, unii-ac80wd7gpf, benzoic acid, ammonium salt 1:1, ac80wd7gpf
PubChem CID 15830
IUPAC Name azanium;benzoate
SMILES C1=CC=C(C=C1)C(=O)[O-].[NH4+]

Specifications

CAS Min % 98.5
Color White
CAS Max % 100.0
Linear Formula CO2NH4C6H5
Beilstein 09, 107
Merck Index 15, 493
Formula Weight 139.15
Percent Purity ≥98.5%
Quantity 250 g
Infrared Spectrum Authentic
Physical Form Crystalline
Chemical Name or Material Benzoic acid ammonium salt, 99%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.