Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoic anhydride, 98%

Catalog No. AC105500025
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 93-97-0 | C14H10O3 | 226.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-97-0
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.23
MDL Number MFCD00003073
InChI Key CHIHQLCVLOXUJW-UHFFFAOYSA-N
Synonym benzoic anhydride, benzoic acid, anhydride, benzoyl anhydride, benzoylbenzoate, benzoic acid anhydride, benzoic acid, 1,1'-anhydride, phenyl anhydride, benzoesaeureanhydrid, unii-9k7x34fov2, phenylcarbonyl benzoate
PubChem CID 7167
ChEBI CHEBI:38815
IUPAC Name benzoyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OC(=O)C2=CC=CC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 38.0°C to 44.0°C
Color White
Density 1.1900g/mL
Boiling Point 360.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula (C6H5CO)2O
Quantity 2.5 kg
Beilstein 09,164
Fieser 01,49; 05,23
Specific Gravity 1.19
Merck Index 15,1094
Solubility Information Solubility in water: 0,01g/L. Other solubilities: soluble in alcohol,chloroform,acetone,benzene,,ethyl acetate,toluene,xylene,ether,glacial,acetic acid,acetic anhydride,moderately soluble in petroleum ether
Formula Weight 226.23
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Benzoic anhydride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and kee
EINECSNumber 202-291-1
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.