Learn More
Benzoin, 99%
Description
Benzoin is used as a flavor, antiseptic and photopolymerization catalyst. It is also used as a raw material in organic syntheses. It serves as a precursor to benzil, which is a photoinitiator. Further, it is used as degassing agent for powder coatings. In addition to this, it is used as an additive and in powder coating to remove the pinhole phenomenon.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Benzoin is used as a flavor, antiseptic and photopolymerization catalyst. It is also used as a raw material in organic syntheses. It serves as a precursor to benzil, which is a photoinitiator. Further, it is used as degassing agent for powder coatings. In addition to this, it is used as an additive and in powder coating to remove the pinhole phenomenon.
Solubility
Soluble in chlorine. Slightly soluble in water, ethanol and ether.
Notes
Moisture sensitive. Incompatible with acids, acid anhydrides, acid chlorides and strong oxidizing agents.
Specifications
Specifications
| Melting Point | 131°C to 136°C |
| Density | 1.31 |
| Boiling Point | 334°C to 344°C |
| Flash Point | 181°C (357°F) |
| Odor | Pleasant |
| Quantity | 1000 g |
| Beilstein | 391839 |
| Merck Index | 14,1093 |
| Solubility Information | Soluble in chlorine. Slightly soluble in water,ethanol and ether. |
| Formula Weight | 212.25 |
| Show More |
Safety and Handling
| EINECSNumber | 204-331-3 |
| RTECSNumber | DI1590000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.