Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoin methyl ether, 97%

Catalog No. p-7080669
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 3524-62-7 | C15H14O2 | 226.275 g/mol

Benzoin methyl ether is used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing , paper varnish and other surface coatings. It is used for anticratering in powder coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzoin methyl ether is used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing , paper varnish and other surface coatings. It is used for anticratering in powder coatings.

Solubility
Soluble in methanol.

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3524-62-7
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.275
MDL Number MFCD00008492
InChI Key BQZJOQXSCSZQPS-UHFFFAOYSA-N
Synonym benzoin methyl ether, methyl benzoin, benzoin methyl ester, o-methylbenzoin, 2-methoxy-2-phenylacetophenone, nisso cure mbo, ethanone, 2-methoxy-1,2-diphenyl, acetophenone, 2-methoxy-2-phenyl, qcu 3, alpha-methoxydeoxybenzoin
PubChem CID 98097
IUPAC Name 2-methoxy-1,2-diphenylethanone
SMILES COC(C1=CC=CC=C1)C(=O)C2=CC=CC=C2

Specifications

Melting Point 46°C to 50°C
Density 1.128
Boiling Point 188°C to 189°C (15 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 2052112
Merck Index 14,1093
Solubility Information Soluble in methanol.
Formula Weight 226.28
Percent Purity 97%
Chemical Name or Material Benzoin methyl ether
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 222-538-7
RTECSNumber AM9350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.