Learn More
Benzophenone, 99%
Description
Benzophenone is widely employed as a photo initiator in UV-curing applications such as inks, imaging and clear coatings in the printing industry. It acts as a UV blocker to prevent photo-degradation of the packaging polymers or its contents. It protects some products such as perfumes and soaps from ultraviolet light. It serves to manufacturers to package the product in clear glass or plastic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Benzophenone is widely employed as a photo initiator in UV-curing applications such as inks, imaging and clear coatings in the printing industry. It acts as a UV blocker to prevent photo-degradation of the packaging polymers or its contents. It protects some products such as perfumes and soaps from ultraviolet light. It serves to manufacturers to package the product in clear glass or plastic.
Solubility
Soluble in ethanol and diethyl ether. Insoluble in water.
Notes
Incompatible with strong oxidizing agents and reducing agents.

Specifications
Specifications
Melting Point | 47°C to 50°C |
Density | 1.111 |
Boiling Point | 305°C |
Flash Point | 138°C (280°F) |
Odor | Pleasant |
Assay Percent Range | 99% |
Quantity | 1000 g |
UN Number | UN3077 |
Beilstein | 1238185 |
Merck Index | 14,1098 |
Show More |
Safety and Handling
Hazard Category | H351-H373-H500 |
Precautionary Statement | P201-P202-P260-P281-P308+P313-P501c |
DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
EINECSNumber | 204-337-6 |
RTECSNumber | DI9950000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.