Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzophenone, 99%

Catalog No. aaa1073930
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Catalog No. AAA1073930

Supplier: Thermo Scientific Chemicals A1073930

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 119-61-9 | C13H10O | 182.222 g/mol

Benzophenone is widely employed as a photo initiator in UV-curing applications such as inks, imaging and clear coatings in the printing industry. It acts as a UV blocker to prevent photo-degradation of the packaging polymers or its contents. It protects some products such as perfumes and soaps from ultraviolet light. It serves to manufacturers to package the product in clear glass or plastic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzophenone is widely employed as a photo initiator in UV-curing applications such as inks, imaging and clear coatings in the printing industry. It acts as a UV blocker to prevent photo-degradation of the packaging polymers or its contents. It protects some products such as perfumes and soaps from ultraviolet light. It serves to manufacturers to package the product in clear glass or plastic.

Solubility
Soluble in ethanol and diethyl ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-61-9
Molecular Formula C13H10O
Molecular Weight (g/mol) 182.222
MDL Number MFCD00003076
InChI Key RWCCWEUUXYIKHB-UHFFFAOYSA-N
Synonym benzophenone, diphenyl ketone, benzoylbenzene, phenyl ketone, methanone, diphenyl, alpha-oxoditane, ketone, diphenyl, benzene, benzoyl, alpha-oxodiphenylmethane, diphenylketone
PubChem CID 3102
ChEBI CHEBI:41308
IUPAC Name diphenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC=C2

Specifications

Melting Point 47°C to 50°C
Density 1.111 g/mL
Boiling Point 305°C
Flash Point 138°C (280°F)
Assay Percent Range 99%
Quantity 250 g
UN Number UN3077
Beilstein 1238185
Merck Index 14,1098
Solubility Information Soluble in ethanol and diethyl ether. Insoluble in water.
Formula Weight 182.22
Percent Purity 99%
Chemical Name or Material Benzophenone
Show More Show Less
Hazard Category H351-H373-H500
Precautionary Statement P201-P202-P260-P281-P308+P313-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 204-337-6
RTECSNumber DI9950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.