Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzophenone, 99+%, pure

Catalog No. ac219680500
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 119-61-9 | C13H10O | 182.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-61-9
Molecular Formula C13H10O
Molecular Weight (g/mol) 182.22
InChI Key RWCCWEUUXYIKHB-UHFFFAOYSA-N
Synonym benzophenone, diphenyl ketone, benzoylbenzene, phenyl ketone, methanone, diphenyl, alpha-oxoditane, ketone, diphenyl, benzene, benzoyl, alpha-oxodiphenylmethane, diphenylketone
PubChem CID 3102
ChEBI CHEBI:41308
IUPAC Name diphenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC=C2

Specifications

Melting Point 47.0°C to 49.0°C
Color White
Boiling Point 305.0°C
Flash Point 143°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)2CO
Quantity 50 g
Beilstein 07,411
Fieser 13,108
Merck Index 15,11
Solubility Information Solubility in water: insoluble. Other solubilities: 1g/7.5mL alcohol,1g/6mL ether,soluble in chloroform,acetone and benzene
Formula Weight 182.22
Percent Purity 99+%
Grade Pure
Physical Form Crystalline Powder or Flakes
Chemical Name or Material Benzophenone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical attention/advice if you feel unwell.
Avoid release to the environment.
Dispose of contents/container to an approved waste disposal plant.
EINECSNumber 204-337-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.