Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzophenone hydrazone, 98+%

Catalog No. AAA122660B
Click to view available options
Quantity:
50 g
250 g
1000 g

CAS: 5350-57-2 | C13H12N2 | 196.253 g/mol

An important compound in organic photochemistry and perfumery as well as in organic synthesis. Used in the antibiotics synthesis of 6-APA and 7-ACA of the carboxyl protecting groups and other organic compounds. Used as organic pigment and medical intermediate. It is used as a photoinitiator of UV-curing applications in inks, adhesive, coatings and optical fiber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An important compound in organic photochemistry and perfumery as well as in organic synthesis. Used in the antibiotics synthesis of 6-APA and 7-ACA of the carboxyl protecting groups and other organic compounds. Used as organic pigment and medical intermediate. It is used as a photoinitiator of UV-curing applications in inks, adhesive, coatings and optical fiber.

Solubility
Soluble in ether, benzene, chloroform and other organic solvents.

Notes
Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances. Keep refrigerated. (Store below 4 C/39 F.)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5350-57-2
Molecular Formula C13H12N2
Molecular Weight (g/mol) 196.253
MDL Number MFCD00007624
InChI Key QYCSNMDOZNUZIT-UHFFFAOYSA-N
Synonym benzophenone hydrazone, diphenylmethylene hydrazine, benzophenonehydrazone, methanone, diphenyl-, hydrazone, diphenylmethanone hydrazone, benzophenone, hydrazone, diphenylmethylidene hydrazine, diphenyl ketone hydrazone, benzophenone hydrozone, nsc 43
PubChem CID 79304
IUPAC Name benzhydrylidenehydrazine
SMILES C1=CC=C(C=C1)C(=NN)C2=CC=CC=C2

Specifications

Melting Point 96°C to 100°C
Density 1.1
Boiling Point 225°C to 230°C (55 mmHg)
Flash Point 146°C (294°F)
Odor Characteristic
Quantity 1000 g
UN Number UN2811
Beilstein 1910177
Solubility Information Soluble in ether,benzene,chloroform and other organic solvents.
Formula Weight 196.25
Percent Purity ≥98%
Chemical Name or Material Benzophenone hydrazone
Show More Show Less
Hazard Category H301-H317
Hazard Statement GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P310-P302+P352-P330-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 226-321-8
RTECSNumber PC4954487
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.