Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Benzoylecgonine Monoclonal Antibody (1P3G2)
GREENER_CHOICE

Catalog No. PIMIB0202
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMIB0202 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMIB0202 Supplier Invitrogen™ Supplier No. MIB0202
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MIB0202 targets Benzoylecgonine in ELISA and RIA applications and shows reactivity with Chemical samples. The MIB0202 immunogen is benzoylecgonine-KLH. MIB0202 was formerly sold as a Seradyn product.

Benzoylecgonine is used as the main pharmaceutical ingredient in the prescription drug Esterom, a topical solution used for the relief of muscle pain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Benzoylecgonine
Applications ELISA, Radioimmune Assays (RIA)
Classification Monoclonal
Clone 1P3G2
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias C16H19NO4
Host Species Mouse
Immunogen Benzoylecgonine-KLH.
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.