Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Benzoylecgonine Monoclonal Antibody (3P1A6)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA514673
Description
MA5-14673 targets Benzoylecgonine in ELISA, IP, and RIA applications and shows reactivity with Chemical samples. The MA5-14673 immunogen is benzoylecgonine-KLH. Product MA514673 is a smaller package size of MIB0201 (formerly sold as a Seradyn product).
Benzoylecgonine is used as the main pharmaceutical ingredient in the prescription drug Esterom, a topical solution used for the relief of muscle pain.
Specifications
Benzoylecgonine | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG1 |
ELISA, Immunoprecipitation, Radioimmune Assays (RIA) | |
3P1A6 | |
Unconjugated | |
C16H19NO4 | |
Benzoylecgonine-KLH. | |
100 μg | |
Primary | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid | |
Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction