Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoylhydrazine, 98%

Catalog No. p-64238
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle

CAS: 613-94-5 | C7H8N2O | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-94-5
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00007596
InChI Key WARCRYXKINZHGQ-UHFFFAOYSA-N
Synonym benzoylhydrazine, benzhydrazide, benzoic hydrazide, benzoyl hydrazine, benzoic acid, hydrazide, benzoic acid hydrazide, benzohydrazine, benzoylhydrazide, benzoyl hydrazide, hydrazine, benzoyl
PubChem CID 11955
ChEBI CHEBI:38454
IUPAC Name benzohydrazide
SMILES NNC(=O)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 113.0°C to 117.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula C6H5CONHNH2
Quantity 100 g
Beilstein 09,319
Solubility Information Solubility in water: soluble
Formula Weight 136.15
Percent Purity 98%
Physical Form Needle-like Crystals or Powder
Chemical Name or Material Benzoylhydrazine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 210-363-9
RTECSNumber DH1575000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.