Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl 2,4-Dihydroxyphenyl Ketone 98.0+%, TCI America™
SDP

Catalog No. B392125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B392125G 25 g
B39215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B392125G Supplier TCI America Supplier No. B392125G

Chemical Identifiers

CAS 3669-41-8
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.25
MDL Number MFCD00043754
InChI Key VFQKAJVKZKHVPD-UHFFFAOYSA-N
Synonym 1-2,4-dihydroxyphenyl-2-phenylethanone, 2',4'-dihydroxy-2-phenylacetophenone, benzyl 2,4-dihydroxyphenyl ketone, 1-2,4-dihydroxyphenyl-2-phenylethan-1-one, 2,4-dihydroxyphenylbenzyl ketone, 2,4-dihydroxydeoxybenzoin, acmc-20aod0, cbmicro_026224, cambridge id 5938997, benzyl2,4-dihydroxyphenylketone
PubChem CID 138005
IUPAC Name 1-(2,4-dihydroxyphenyl)-2-phenylethan-1-one
SMILES OC1=CC=C(C(=O)CC2=CC=CC=C2)C(O)=C1

Specifications

Melting Point 113°C
Boiling Point 225°C
Quantity 25 g
Formula Weight 228.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl 2,4-Dihydroxyphenyl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.