Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadmium chloride, 99.99%, (trace metal basis)
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
Specifications
Specifications
| Boiling Point | 960.0°C |
| Melting Point | 568.0°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 25 g |
| Assay Percent Range | 99.99% |
| Linear Formula | CdCl2 |
| Fieser | 04,485 |
| Merck Index | 15, 1622 |
| Solubility Information | Solubility in water: 1400g/L (20°C). Other solubilities: soluble in acetone, slightly soluble in methanol and ethanol, practically insoluble in ether |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause genetic defects. Causes damage to organs through prolonged or repeated exposure. Fatal if inhaled. May cause cancer. May damage fertility. May damage the unborn child. Toxic if swallowed |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Obtain special instructions before use. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF S |
| EINECSNumber | 233-296-7 |
| RTECSNumber | EV0175000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction