Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl 3-oxopiperazine-1-carboxylate, 97%, Thermo Scientific™

Catalog No. km07871de
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 78818-15-2
Molecular Formula C12H14N2O3
Molecular Weight (g/mol) 234.255
InChI Key BAHFPJFBMJTOPU-UHFFFAOYSA-N
Synonym 4-benzyloxycarbonylpiperazin-2-one, 1-cbz-3-oxopiperazine, 4-benzyloxycarbonyl-2-piperazinone, 4-cbz-piperazinone, 1-z-3-oxopiperazine, 4-cbz-piperazin-2-one, 1-piperazinecarboxylic acid, 3-oxo-, phenylmethyl ester, 3-oxopiperazine-1-carboxylic acid benzyl ester, benzyl 3-oxotetrahydro-1 2h-pyrazinecarboxylate, phenylmethyl 3-oxopiperazinecarboxylate
PubChem CID 736777
IUPAC Name benzyl 3-oxopiperazine-1-carboxylate
SMILES C1CN(CC(=O)N1)C(=O)OCC2=CC=CC=C2

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 234.25
Percent Purity 97%
Chemical Name or Material Benzyl 3-oxopiperazine-1-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.