Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl 4-Hydroxy-1-piperidinecarboxylate 98.0+%, TCI America™
SDP

Catalog No. B486925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B486925G 25 g
B48695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B486925G Supplier TCI America Supplier No. B486925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 95798-23-5
Molecular Formula C13H17NO3
Molecular Weight (g/mol) 235.28
MDL Number MFCD01863722
InChI Key JKIUUDJOCYHIGY-UHFFFAOYSA-N
Synonym benzyl 4-hydroxy-1-piperidinecarboxylate, 1-cbz-4-hydroxypiperidine, n-cbz-4-hydroxy-1-piperidine, benzyl 4-hydroxytetrahydro-1 2h-pyridinecarboxylate, n-benzyloxycarbonyl-4-hydroxypiperidine, 1-piperidinecarboxylic acid, 4-hydroxy-, phenylmethyl ester, n-cbz-4-hydroxypiperidine, n-benzyloxycarbonyl piperidin-4-ol, 4-hydroxypiperidine, n-cbz protected, 4-hydroxy-piperidine-1-carboxylic acid benzyl ester
PubChem CID 2794620
IUPAC Name benzyl 4-hydroxypiperidine-1-carboxylate
SMILES OC1CCN(CC1)C(=O)OCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 167°C
Quantity 25 g
Formula Weight 235.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl 4-Hydroxy-1-piperidinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.