Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl acetate, 99%

Catalog No. aab2347236
Click to view available options
Quantity:
500 g
2500 g

CAS: 140-11-4 | C9H10O2 | 150.177 g/mol

Benzyl acetate can be used as a high boiling point solvent in coatings, like ink coating, binding agent and paint remover. It is used in soap and other chemical essences and has the effect of promote in floral and fantasy essences in jasmine, white orchid, fragrant plantain lily, gekkakou, narcissus and other essences. It is a plasticizer for ionophore membranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl acetate can be used as a high boiling point solvent in coatings, like ink coating, binding agent and paint remover. It is used in soap and other chemical essences and has the effect of promote in floral and fantasy essences in jasmine, white orchid, fragrant plantain lily, gekkakou, narcissus and other essences. It is a plasticizer for ionophore membranes.

Solubility
Soluble in water 3.1 g/L.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-11-4
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00008712
InChI Key QUKGYYKBILRGFE-UHFFFAOYSA-N
Synonym acetic acid benzyl ester, benzyl ethanoate, acetic acid, phenylmethyl ester, phenylmethyl acetate, acetic acid, benzyl ester, alpha-acetoxytoluene, phenylmethyl ethanoate, acetoxymethyl benzene, benzylester kyseliny octove, caswell no. 081ea
PubChem CID 8785
ChEBI CHEBI:52051
IUPAC Name benzyl acetate
SMILES CC(=O)OCC1=CC=CC=C1

Specifications

Melting Point °C to 51°C
Density 1.053
Boiling Point 205°C to 207°C
Flash Point 95°C (203°F)
Assay Percent Range 99+%
Refractive Index 1.502
Quantity 500 g
Beilstein 1908121
Merck Index 14,1123
Solubility Information Soluble in water 3.1g/L.
Formula Weight 150.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Benzyl acetate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-399-7
RTECSNumber AF5075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.