Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl alcohol, 98+%, Extra Dry, AcroSeal™

Catalog No. p-3756889
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle

CAS: 100-51-6 | C7H8O | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00004599,MFCD03792087
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
Synonym benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
PubChem CID 244
ChEBI CHEBI:17987
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -15.0°C
Color Colorless
Density 1.0440g/mL
Boiling Point 205.0°C
Flash Point 96°C
Assay Percent Range 98% min. (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.5380 to 1.5410
Linear Formula C6H5CH2OH
Quantity 100 mL
Residue after Evaporation 0.05% max.
Specific Gravity 1.044
Merck Index 15, 1127
Solubility Information Solubility in water: 40g/L (20°C). Other solubilities: miscible with alcohol,ether,acetone,chloroform,,dmf and methylene chloride
Viscosity 6.6 mPa.s (20°C)
Formula Weight 108.14
Percent Purity 98+%
Grade Extra Dry
Physical Form Liquid
Chemical Name or Material Benzyl alcohol, Extra Dry
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/phys
EINECSNumber 202-859-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.