Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl alcohol, ACS, 99+%

Catalog No. p-7051447
Click to view available options
Quantity:
250 mL
1 L
4 L
4 x 1 L

Catalog No. AA41218AK

Supplier: Thermo Scientific Chemicals 041218AK

Only null left
Add to Cart

CAS: 100-51-6 | C7H8O | 108.14 g/mol

Benzyl alcohol is widely used as a solvent for the dielectrophoretic reconfiguration of nanowires, inks, paints, lacquers and epoxy resin coatings and as a precursor to a variety of esters used in soaps, perfumes and flavoring. It is employed as a local anesthetic which reduces the pain associated with lidocaine injection. It has a various applications in baby products, bath products, soaps and detergents, eye makeup, blushers, cleansing products, make up products as well as hair, nail and skin care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00004599,MFCD03792087
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
Synonym benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
PubChem CID 244
ChEBI CHEBI:17987
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1

Specifications

Melting Point -15°C
Density 1.045 g/mL
Boiling Point 205°C
Flash Point 100°C (212°F)
Odor Aromatic
Refractive Index 1.54
Quantity 250 mL
Beilstein 878307
Merck Index 14,1124
Solubility Information Misicible with water,benzene,methanol,chloroform,ethanol,acetone and ether.
Formula Weight 108.14
Percent Purity ≥99%
Grade ACS
Physical Form Liquid
Chemical Name or Material Benzyl alcohol
Show More Show Less
Hazard Category H302+H332-H319
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P314-P330-P501c
EINECSNumber 202-859-9
RTECSNumber DN3150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.