Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl alcohol, for analysis, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass bottle
100-51-6
C7H8O
108.14
MFCD00004599
WVDDGKGOMKODPV-UHFFFAOYSA-N
benzyl alcohol,benzenemethanol,phenylcarbinol,benzoyl alcohol,hydroxytoluene,benzenecarbinol,phenylmethyl alcohol,alpha-toluenol,hydroxymethyl benzene,benzylalcohol
244
CHEBI:17987
phenylmethanol
C1=CC=C(C=C1)CO
100-51-6
C7H8O
108.14
MFCD00004599
WVDDGKGOMKODPV-UHFFFAOYSA-N
benzyl alcohol,benzenemethanol,phenylcarbinol,benzoyl alcohol,hydroxytoluene,benzenecarbinol,phenylmethyl alcohol,alpha-toluenol,hydroxymethyl benzene,benzylalcohol
244
CHEBI:17987
phenylmethanol
C1=CC=C(C=C1)CO
For analysis
Viscosity 6.6 mPa.s (20°C)
Acidity 0.001 meq/g max.
Water 0.1% max. (K.F.)
Refractive Index 1.5380 to 1.5410
Specific Gravity 1.044
Quantity 1L
Alkalinity 0.002 meq/g max.
Packaging Glass bottle
Infrared Spectrum Authentic
Flash Point 96°C
Boiling Point 205.0°C
Color Undesignated
Density 1.0440g/mL
Melting Point -15.0°C
Linear Formula C6H5CH2OH
Merck Index 15, 1127
Solubility Information Solubility in water: 40g/L (20°C). Other solubilities: miscible with alcohol,ether,acetone,chloroform,,dmf and methylene chloride
Formula Weight 108.14
Physical Form Liquid
Percent Purity ≥99.5%
Grade Analysis
Chemical Name or Material Benzyl alcohol
Show More Show Less

GHS H Statement
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/phys

GHS Signal Word: Warning

EINECSNumber : 202-859-9