Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl alcohol, Analytical Standard, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7193413 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
5 mL
25 mL

Benzyl alcohol belongs to the aromatic alcohol group of compounds widely utilized in cosmetics and preservatives. It is also used as a solvent and viscosity-reducing agent.

TRUSTED_SUSTAINABILITY

Specifications

CAS 100-51-6
Vapor Pressure 3.75 mmHg (77°C)
Vapor Density 3.7 (vs. air)
Density 1.045 g/mL (at 25°C)
Linear Formula C6H5CH2OH
MDL Number MFCD00004599
Melting Point -16°C to +13°C
Boiling Point 203°C to 205°C
Synonym Benzenemethanol
Quantity 5 mL
Formula Weight 108.14
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.