Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Alcohol (NF/EP/BP), Fisher Chemical

Catalog No. A3924 Shop All Fisherbrand Products
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00004599,MFCD03792087
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
Synonym benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
PubChem CID 244
ChEBI CHEBI:17987
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1

Specifications

Acidity Pass Test (BP/EP/NF)
Melting Point -15°C
Color Yellow
Density 1.044g/cm³
Boiling Point 205°C
Packaging Glass Bottle
Refractive Index 1.538 to 1.541 (at 20°C) (BP/EP/NF)
Linear Formula C6H5CH2OH
Quantity 4 L
Residual Solvents Meets Requirements (BP/EP/NF)
Vapor Density 3.72g/mol
Vapor Pressure 0.13mbar at 20°C
Formula Weight 108.14
Percent Purity 98.0 to 100.5% (EP)
Grade BP/EP/NF
Physical Form Liquid
Chemical Name or Material Benzyl Alcohol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.