Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl azide, 94%

Catalog No. AA3263518
Click to view available options
Quantity:
2 g
10 g
50 g

CAS: 622-79-7 | C7H7N3 | 133.154 g/mol

Benzyl azide is used in the synthesis of 1,2,3-triazole derivatives. It serves as a reagent used to introduce a PhCH2N group into a molecule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-79-7
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.154
MDL Number MFCD00013836
InChI Key UDLLFLQFQMACJB-UHFFFAOYSA-N
Synonym azidomethyl benzene, benzyl azide, benzylazide, benzene, azidomethyl, alpha-azidotoluene, toluene, .alpha.-azido, unii-hfd57z7j9j, .alpha.-azidotoluene, ccris 8029
PubChem CID 12152
IUPAC Name azidomethylbenzene
SMILES C1=CC=C(C=C1)CN=[N+]=[N-]

Specifications

Color Brown to Colorless
Density 1.065
Boiling Point 82°C to 85°C (16 mmHg)
Flash Point 43°C (110°F)
Refractive Index 1.5341
Quantity 50 g
UN Number UN1993
Solubility Information Insoluble in waterMiscible with ethanol and diethyl ether; Immiscible with water.
Formula Weight 133.15
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Benzyl azide
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226;
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
RTECSNumber XS6650000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.