Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl bromide, 98%

Catalog No. AC105870010
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Glass bottle

CAS: 100-39-0 | C7H7Br | 171.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-39-0
MDL Number MFCD00000172
InChI Key AGEZXYOZHKGVCM-UHFFFAOYSA-N
Synonym benzyl bromide, bromomethyl benzene, alpha-bromotoluene, bromophenylmethane, benzene, bromomethyl, phenylmethyl bromide, 1-bromotoluene, benzylbromide, cyclite
PubChem CID 7498
ChEBI CHEBI:59858
IUPAC Name bromomethylbenzene
SMILES C1=CC=C(C=C1)CBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -2°C
Color Colorless to Yellow
Density 1.4300g/mL
Boiling Point 198.0°C to 199.0°C
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H7Br
Refractive Index 1.5742 to 1.5762
Linear Formula C6H5CH2Br
Quantity 1 L
Beilstein 05, 306
Fieser 05,25; 13,51
Specific Gravity 1.43
Merck Index 15, 1131
Solubility Information Solubility in water: 436 mg/l (25°C). Other solubilities: slowly decomposes, soluble in alcohol, benzene and ether
Molecular Weight (g/mol) 171.04
Formula Weight 171.04
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Benzyl bromide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF
EINECSNumber 202-847-3
RTECSNumber XS7965000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.