Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl carbamate, 99%

Catalog No. AAA1156922
Click to view available options
Quantity:
25 g
100 g
250 g

CAS: 621-84-1 | C8H9NO2 | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-84-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007965
InChI Key PUJDIJCNWFYVJX-UHFFFAOYSA-N
Synonym carbamic acid benzyl ester, carbamic acid, phenylmethyl ester, o-benzyl carbamate, ccris 5070, phenylmethyl aminooate, carbamic acid, benzyl ester, benzyl-carbamate, benzyloxycarbonylamine, cbznh2, pubchem12421
PubChem CID 12136
IUPAC Name benzyl carbamate
SMILES NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 85°C to 89°C
Beilstein 1865635
Quantity 100 g
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material Benzyl carbamate
Hazard Category H332-H335
Precautionary Statement P261-P271-P304+P340-P312-P501c
EINECSNumber 210-710-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.