Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Carbazate 98.0+%, TCI America™
SDP

Catalog No. C156425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C156425G 25 g
C15645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C156425G Supplier TCI America Supplier No. C156425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5331-43-1
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00041890
InChI Key RXUBZLMIGSAPEJ-UHFFFAOYSA-N
Synonym benzyl carbazate, benzyl hydrazinecarboxylate, hydrazinecarboxylic acid, phenylmethyl ester, carbobenzoxyhydrazide, carbazic acid, benzyl ester, benzyloxycarbonyl hydrazide, benzylcarbazate, benzyloxy carbonyl hydrazine, n-cbz-hydrazine, unii-h7gm2h55tc
PubChem CID 79242
IUPAC Name benzyl N-aminocarbamate
SMILES C1=CC=C(C=C1)COC(=O)NN

Specifications

Melting Point 69°C
Color White-Yellow
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl Carbazate
EINECSNumber (3)-4377
RTECSNumber MV1724950
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.