Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl chloride, 99%, pure, stabilized

Catalog No. AC405090010
Click to view available options
Quantity:
1 kg
10 kg
Packaging:
Glass Bottle
Plastic drum

CAS: 100-44-7 | C7H7Cl | 126.583 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-44-7, 75-56-9
MDL Number MFCD00000889
InChI Key KCXMKQUNVWSEMD-UHFFFAOYSA-N
Synonym benzyl chloride, chloromethyl benzene, benzylchloride, alpha-chlorotoluene, benzylchlorid, chlorophenylmethane, benzene, chloromethyl, chlorure de benzyle, tolyl chloride, alpha-chlortoluol
PubChem CID 7503
ChEBI CHEBI:615597
IUPAC Name chloromethylbenzene
SMILES C1=CC=C(C=C1)CCl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Molecular Formula C7H7Cl
Refractive Index 1.5370 to 1.5390 (20°C, 589nm)
Linear Formula C6H5CH2Cl
Quantity 1 kg
Stabilizer 0.25% propylene oxide
Molecular Weight (g/mol) 126.583
Formula Weight 126.59
Percent Purity ≥98.5%
Grade Pure
Physical Form Liquid
Chemical Name or Material Benzyl chloride, Stabilized, Pure
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Fatal if inhaled.
May cause cancer.
May cause respiratory irritation.
Causes skin irritation.
May cause genetic defects.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.