Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl chloride, 99%, stab.

Catalog No. aaa1248136
Click to view available options
Quantity:
10,000 g
2500 g
500 g

Catalog No. AAA1248136

Supplier: Thermo Scientific Chemicals A1248136

Only null left
Add to Cart

CAS: 100-44-7 | C7H7Cl | 126.583 g/mol

Benzyl Chloride is used as an intermediate for manufacturing of benzyl alcohol, benzoic acid and benzyl cyanide. It is also used to prepare benzyl esters, which are used as plasticizer, flavorants and perfumes. It acts as an intermediate in the preparation of phenylacetic acid. In organic synthesis, it is used as an alkylating agent and utilized for the introduction of the benzyl protecting group for alcohols. Further, it is involved in the preparation of quaternary ammonium salts by reaction with tertiary amines. In addition to this, it is used to prepare Grignard reagent by reacting with metallic magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl Chloride is used as an intermediate for manufacturing of benzyl alcohol, benzoic acid and benzyl cyanide. It is also used to prepare benzyl esters, which are used as plasticizer, flavorants and perfumes. It acts as an intermediate in the preparation of phenylacetic acid. In organic synthesis, it is used as an alkylating agent and utilized for the introduction of the benzyl protecting group for alcohols. Further, it is involved in the preparation of quaternary ammonium salts by reaction with tertiary amines. In addition to this, it is used to prepare Grignard reagent by reacting with metallic magnesium.

Solubility
Miscible with alcohol, ether. Insoluble in waterMiscible with alcohol, ether and chloroform.

Notes
Light and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with common metals, lead,oxidizing agents, iron, iron salts, brass and aluminum
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-44-7
Molecular Formula C7H7Cl
Molecular Weight (g/mol) 126.583
MDL Number MFCD00000889
InChI Key KCXMKQUNVWSEMD-UHFFFAOYSA-N
Synonym benzyl chloride, chloromethyl benzene, benzylchloride, alpha-chlorotoluene, benzylchlorid, chlorophenylmethane, benzene, chloromethyl, chlorure de benzyle, tolyl chloride, alpha-chlortoluol
PubChem CID 7503
ChEBI CHEBI:615597
IUPAC Name chloromethylbenzene
SMILES C1=CC=C(C=C1)CCl

Specifications

Melting Point - 43°C
Density 1.099
Boiling Point 178°C to 180°C
Flash Point 73°C (163°F)
Assay Percent Range 99%, Stabilized
Refractive Index 1.538
Quantity 500 g
UN Number UN1738
Beilstein 471308
Merck Index 14,1129
Solubility Information Miscible with alcohol,ether; Insoluble in waterMiscible with alcohol,ether and chloroform.
Formula Weight 126.59
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Benzyl Chloride, stabilized
Show More Show Less
Hazard Category H227-H290-H302-H315-H317-H318-H331-H335-H336-H340-H350-H373
Hazard Statement GHS H Statement
H301-H330-H350-H373-H314-H318-H227-H335
Precautionary Statement P201-P202-P210-P234-P235-P260-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P311-P330-P333+P313-P362-P370+P378q-P390-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: BENZYL CHLORIDE
EINECSNumber 202-853-6
RTECSNumber XS8925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.