Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Ether 99%, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass bottle
103-50-4
C14H14O
198.26
MFCD00004780
MHDVGSVTJDSBDK-UHFFFAOYSA-N
dibenzyl ether,benzyl ether,benzyl oxide,dibenzylether,oxybis methylene dibenzene,plastikator ba,ba plasticizer,benzylether,ether, dibenzyl,plasticator ba
7657
CHEBI:87411
phenylmethoxymethylbenzene
C1=CC=C(C=C1)COCC2=CC=CC=C2
103-50-4
C14H14O
198.26
MFCD00004780
MHDVGSVTJDSBDK-UHFFFAOYSA-N
dibenzyl ether,benzyl ether,benzyl oxide,dibenzylether,oxybis methylene dibenzene,plastikator ba,ba plasticizer,benzylether,ether, dibenzyl,plasticator ba
7657
CHEBI:87411
phenylmethoxymethylbenzene
C1=CC=C(C=C1)COCC2=CC=CC=C2
Melting Point 1.5°C to 3.5°C
Flash Point 135°C
Refractive Index 1.5610 to 1.5630
Packaging Glass bottle
Infrared Spectrum Authentic
Specific Gravity 1.04
Boiling Point 298.0°C
Quantity 1L
Density 1.0400g/mL
Color Colorless to Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (C6H5CH2)2O
Beilstein 06, 434
Merck Index 15, 1135
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,acetone and chloroform
Formula Weight 198.26
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Benzyl ether, 99%
Show More Show Less

EINECSNumber : 203-118-2

RTECSNumber : DQ6125000

TSCA : TSCA