Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl isocyanate, 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
3173-56-6
C8H7NO
133.15
MFCD00009701
YDNLNVZZTACNJX-UHFFFAOYSA-N
benzyl isocyanate,isocyanatomethyl benzene,benzylisocyanate,isocyanic acid benzyl ester,benzene, isocyanatomethyl,1-isocyanatomethyl benzene,phenylmethyl isocyanate,unii-8i2li357gq,phenylmethanisocyanate
76639
isocyanatomethylbenzene
C1=CC=C(C=C1)CN=C=O
3173-56-6
C8H7NO
133.15
MFCD00009701
YDNLNVZZTACNJX-UHFFFAOYSA-N
benzyl isocyanate,isocyanatomethyl benzene,benzylisocyanate,isocyanic acid benzyl ester,benzene, isocyanatomethyl,1-isocyanatomethyl benzene,phenylmethyl isocyanate,unii-8i2li357gq,phenylmethanisocyanate
76639
isocyanatomethylbenzene
C1=CC=C(C=C1)CN=C=O
Specific Gravity 1.07
Boiling Point 101°C to 104°C (33.0mmHg)
Density 1.0700g/mL
Flash Point 45°C
Refractive Index 1.5241 to 1.5261
Quantity 25g
Infrared Spectrum Authentic
Color Colorless to Yellow
Packaging Glass bottle
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula C6H5CH2NCO
Beilstein 12, 1059
Fieser 13,30; 01,1005
Formula Weight 133.15
Physical Form Liquid
Percent Purity 99+%
Chemical Name or Material Benzyl isocyanate, 99+%
Show More Show Less

GHS H Statement:
Toxic if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Toxic in contact with skin.
Toxic if swallowed.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 221-640-9