Learn More
Titanium(IV) oxide, 98+%, anatase powder
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 2900.0°C |
| Melting Point | 1855.0°C |
| Color | White |
| pH | 7.3 to 8.3 (aq. suspension) |
| Physical Form | Powder |
| Quantity | 1 kg |
| Assay Percent Range | 98% min |
| Linear Formula | TiO2 |
| Merck Index | 15, 9628 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in hot concentrated h2so4 and hf |
| Show More |
Frequently Asked Questions (FAQs)
Rutile and anatase are polymorphs of TiO2, which are both naturally occurring. Rutile is a source for titanium smelting and is used in the semiconductor industry to produce dye-based solar cells (Grätzel cells). Anatase shows a high photocatalytic activity, which limits the usability as a pigment in polymers, but also opens certain applications as a catalyst. Anatase has a higher photocatalytic activity than rutile but is thermodynamically less stable.
Safety and Handling
| EINECSNumber | 236-675-5 |
| RTECSNumber | XR2275000 |
| TSCA | TSCA |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.