Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Benzyl Isothiocyanate 98.0+%, TCI America™

Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Boiling Point | 243°C |
Quantity | 25 g |
UN Number | 3082 |
Formula Weight | 149.21 |
Percent Purity | ≥98.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | Benzyl Isothiocyanate |
Safety and Handling
EINECSNumber | (3)-3243 |
RTECSNumber | NX8250000 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction