Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl mercaptan, 99%

Catalog No. AAB2008122
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 100-53-8 | C7H8S | 124.20 g/mol

A common laboratory alkylthiol, which can be used as a source of thiol function. It also finds it uses as an odorant, synthetic flavor, and as a bacteriostat.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A common laboratory alkylthiol, which can be used as a source of thiol function. It also finds it uses as an odorant, synthetic flavor, and as a bacteriostat.

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-53-8
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.20
MDL Number MFCD00004867,MFCD00801588,MFCD01863867
InChI Key UENWRTRMUIOCKN-UHFFFAOYSA-N
Synonym benzyl mercaptan, benzenemethanethiol, benzylthiol, alpha-toluenethiol, thiobenzyl alcohol, benzyl hydrosulfide, alpha-toluolthiol, benzylhydrosulfide, phenylmethyl mercaptan, benzylmercaptan
PubChem CID 7509
IUPAC Name phenylmethanethiol
SMILES SCC1=CC=CC=C1

Specifications

Melting Point °C to 30°C
Density 1.053
Boiling Point 194°C to 195°C
Flash Point 70°C (158°F)
Assay Percent Range 0.99
Refractive Index 1.576
Quantity 100 g
UN Number UN2810
Beilstein 605864
Sensitivity Air Sensitive
Merck Index 14,9322
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 124.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Benzyl mercaptan
Show More Show Less
Hazard Category H227-H302-H319-H330
Hazard Statement GHS H Statement
H330-H227-H302
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P284-P301+P312-P304+P340-P305+P351+P338-P310-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 202-862-5
RTECSNumber XT8650000
TSCA Yes
storageNote1 Air Sensitive
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.