Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl mercaptan, 99%

Catalog No. AC421265000
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 100-53-8 | C7H8S | 124.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-53-8
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.20
MDL Number MFCD00004867,MFCD00801588,MFCD01863867
InChI Key UENWRTRMUIOCKN-UHFFFAOYSA-N
Synonym benzyl mercaptan, benzenemethanethiol, benzylthiol, alpha-toluenethiol, thiobenzyl alcohol, benzyl hydrosulfide, alpha-toluolthiol, benzylhydrosulfide, phenylmethyl mercaptan, benzylmercaptan
PubChem CID 7509
IUPAC Name phenylmethanethiol
SMILES SCC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -29°C
Color Colorless to Yellow
Density 1.0580g/mL
Boiling Point 195°C
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5735 to 1.5775
Linear Formula C6H5CH2SH
Quantity 500 g
Beilstein 06, IV, 2632
Specific Gravity 1.058
Merck Index 15, 9475
Solubility Information Solubility in water: insoluble
Formula Weight 124.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Benzyl mercaptan
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: rinse mouth.
Do NO
EINECSNumber 202-862-5
Recommended Storage Product darkens during storage

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.