Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl mercaptan, polymer supported, 1% cross-linked, 100-200 mesh, 2.0-4.0 mmol/g on poly(styrene-divinylbenzene)

Catalog No. p-7059358
Click to view available options
Quantity:
2 g
10 g
50 g

CAS: 100-53-8 | C7H8S | 124.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-53-8
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.20
MDL Number MFCD00004867,MFCD00801588,MFCD01863867
InChI Key UENWRTRMUIOCKN-UHFFFAOYSA-N
Synonym benzyl mercaptan, benzenemethanethiol, benzylthiol, alpha-toluenethiol, thiobenzyl alcohol, benzyl hydrosulfide, alpha-toluolthiol, benzylhydrosulfide, phenylmethyl mercaptan, benzylmercaptan
PubChem CID 7509
IUPAC Name phenylmethanethiol
SMILES SCC1=CC=CC=C1

Specifications

Color Pink to White
Mesh Size 100 to 200 Mesh
Quantity 2 g
Physical Form Beads
Chemical Name or Material Benzyl mercaptan, Polymer supported, 1% cross-linked, 2.0 to 4.0 mmol/g on poly(styrene-divinylbenzene)
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.