Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl n-butyl phthalate, 98%

Catalog No. AAB247690E
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 85-68-7 | C19H20O4 | 312.365 g/mol

Benzyl n-butyl phthalate is used as a plasticizer for vinyl foams. It is also used in floor tiles, in traffic cones, food conveyor belts and artificial leather. Further, it acts as an organic intermediate. In addition to this, it is used as a perfume fixative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl n-butyl phthalate is used as a plasticizer for vinyl foams. It is also used in floor tiles, in traffic cones, food conveyor belts and artificial leather. Further, it acts as an organic intermediate. In addition to this, it is used as a perfume fixative.

Solubility
Slightly miscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, alkalis and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-68-7
Molecular Formula C19H20O4
Molecular Weight (g/mol) 312.365
MDL Number MFCD00009440
InChI Key IRIAEXORFWYRCZ-UHFFFAOYSA-N
Synonym benzyl butyl phthalate, butyl benzyl phthalate, sicol, palatinol bb, unimoll bb, santicizer 160, butylbenzyl phthalate, n-butyl benzyl phthalate, sicol 160, benzyl n-butyl phthalate
PubChem CID 2347
ChEBI CHEBI:34595
IUPAC Name 2-O-benzyl 1-O-butyl benzene-1,2-dicarboxylate
SMILES CCCCOC(=O)C1=CC=CC=C1C(=O)OCC2=CC=CC=C2

Specifications

Density 1.1
Boiling Point 370°C
Flash Point >110°C (230°F)
Refractive Index 1.54
Quantity 2500 g
UN Number UN3082
Solubility Information Slightly miscible with water.
Formula Weight 312.37
Percent Purity 98%
Chemical Name or Material Benzyl n-butyl phthalate
Hazard Category H319-H360
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 201-622-7
TSCA Yes

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.