Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl phenyl ether, 97%

Catalog No. p-7026514
Click to view available options
Quantity:
5 g
25 g
100 g

Catalog No. AAB2253906

Supplier: Thermo Scientific Chemicals B2253906

Only null left
Add to Cart

CAS: 946-80-5 | C13H12O | 184.238 g/mol

Benzyl phenyl ether reacts with aluminum bromide in chlorobenzene solution to afford a mixture of phenol, o-benzyl phenol and dichlorodiphenylmethane. It is a useful as model compound in catalytic chemistry to represent the a-O4 ether bond in lignin and coal. It contains a weak ether bond of 234kJ/mol and belongs to the most thermo-labile compounds in lignin and low rank coal. Influence of alkali carbonates, common additives in biomass conversion, on the reaction pathways of BPE in superheated water has been reported. Cesium-exchanged heteropolyacid catalyzed decomposition of benzyl phenyl ether to aromatics has been investigated. Photo-Claisen rearrangements of benzyl phenyl ether was investigated in cation-exchanged Y zeolites and polyethylenes of differing crystallinities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl phenyl ether reacts with aluminum bromide in chlorobenzene solution to afford a mixture of phenol, o-benzyl phenol and dichlorodiphenylmethane. It is a useful as model compound in catalytic chemistry to represent the a-O4 ether bond in lignin and coal. It contains a weak ether bond of 234kJ/mol and belongs to the most thermo-labile compounds in lignin and low rank coal. Influence of alkali carbonates, common additives in biomass conversion, on the reaction pathways of BPE in superheated water has been reported. Cesium-exchanged heteropolyacid catalyzed decomposition of benzyl phenyl ether to aromatics has been investigated. Photo-Claisen rearrangements of benzyl phenyl ether was investigated in cation-exchanged Y zeolites and polyethylenes of differing crystallinities.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 946-80-5
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.238
MDL Number MFCD00020660
InChI Key BOTNYLSAWDQNEX-UHFFFAOYSA-N
Synonym benzyl phenyl ether, benzyloxy benzene, phenyl benzyl ether, benzene, phenoxymethyl, ether, benzyl phenyl, benzylphenylether, benzyloxy-benzene, benzyloxybenzene, unii-bue863n0l8, .alpha.-phenylanisole
PubChem CID 70352
IUPAC Name phenoxymethylbenzene
SMILES C1=CC=C(C=C1)COC2=CC=CC=C2

Specifications

Melting Point 37°C to 40°C
Boiling Point 286°C to 287°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 2045713
Solubility Information Insoluble in water.
Formula Weight 184.24
Percent Purity 97%
Chemical Name or Material Benzyl phenyl ether
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.