Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzylacetophenone, MP Biomedicals

Catalog No. ICN224314
Click to view available options
Quantity:
10 g

Chemical Identifiers

CAS 1083-30-3
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.276
InChI Key QGGZBXOADPVUPN-UHFFFAOYSA-N
Synonym dihydrochalcone, 3-phenylpropiophenone, benzylacetophenone, hydrochalcone, 1,3-diphenyl-1-propanone, beta-phenylpropiophenone, hydrocinnamophenone, 1,3-diphenyl-1-oxopropane, phenethyl phenyl ketone, phenyl phenethyl ketone
PubChem CID 64802
ChEBI CHEBI:71231
IUPAC Name 1,3-diphenylpropan-1-one
SMILES C1=CC=C(C=C1)CCC(=O)C2=CC=CC=C2

Specifications

Quantity 10 g
Formula Weight 210.29
Chemical Name or Material Benzylacetophenone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.