Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzylamine, 98+%

Catalog No. p-7081150
Change view
Click to view available options
Quantity:
250 g
500 g
1000 g
5000 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1099730 250 g
AAA1099736 500 g
AAA109970B 1000 g
AAA109970I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1099730 Supplier Thermo Scientific Chemicals Supplier No. A1099730
Only null left
Add to Cart

CAS: 100-46-9 | C7H9N | 107.156 g/mol

Benzylamine is a valuable intermediate for various applications and a building block for chemical synthesis used in pharmaceuticals and crop protection agents. It finds application in the coating industry. It is involved as a carrier electrolyte for separation of alkali, alkaline earth and ammonium cations in water samples by capillary electrophoresis with indirect UV detection. It is also used in synthesis of cross-linked porous copolymer supports based on N-(p-vinylbenzoyl)-2-methylalanine, styrene and divinylbenzene. In the textile industry, it is used in colored dyes. It is often used for medicinal purposes in topical creams and antifungal solutions as well as in vitamins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzylamine is a valuable intermediate for various applications and a building block for chemical synthesis used in pharmaceuticals and crop protection agents. It finds application in the coating industry. It is involved as a carrier electrolyte for separation of alkali, alkaline earth and ammonium cations in water samples by capillary electrophoresis with indirect UV detection. It is also used in synthesis of cross-linked porous copolymer supports based on N-(p-vinylbenzoyl)-2-methylalanine, styrene and divinylbenzene. In the textile industry, it is used in colored dyes. It is often used for medicinal purposes in topical creams and antifungal solutions as well as in vitamins.

Solubility
Miscible with water, ethanol, diethyl ether, acetone, benzene and chloroform.

Notes
Air sensitive. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-46-9
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00008106
InChI Key WGQKYBSKWIADBV-UHFFFAOYSA-N
Synonym benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine
PubChem CID 7504
ChEBI CHEBI:40538
IUPAC Name phenylmethanamine
SMILES C1=CC=C(C=C1)CN

Specifications

Melting Point -43°C
Density 0.982
Boiling Point 184°C to 185°C
Flash Point 72°C (162°F)
Refractive Index 1.5435
Quantity 250 g
UN Number UN2735
Beilstein 741984
Sensitivity Air Sensitive
Merck Index 14,1125
Solubility Information Miscible with water,ethanol,diethyl ether,acetone,benzene and chloroform.
Formula Weight 107.16
Percent Purity ≥98%
Chemical Name or Material Benzylamine
Show More Show Less
Hazard Category H227-H302+H312-H314-H335
Hazard Statement GHS H Statement
H314-H302-H312-H227
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
Precautionary Statement P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P363-P374-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 202-854-1
RTECSNumber DP1488500
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.