CAS RN 100-46-9
Benzylamine, 99.5+%, AcroSeal™
CAS: 100-46-9 Molecular Formula: C7H9N Molecular Weight (g/mol): 107.15 MDL Number: MFCD00008106 InChI Key: WGQKYBSKWIADBV-UHFFFAOYSA-N Synonym: benzylamine,benzenemethanamine,monobenzylamine,alpha-aminotoluene,phenylmethyl amine,aminomethyl benzene,1-phenylmethanamine,moringine,n-benzylamine,phenylmethylamine PubChem CID: 7504 ChEBI: CHEBI:40538 IUPAC Name: phenylmethanamine SMILES: C1=CC=C(C=C1)CN
Benzylamine, 99%, pure
CAS: 100-46-9 Molecular Formula: C7H9N Molecular Weight (g/mol): 107.15 MDL Number: MFCD00008106 InChI Key: WGQKYBSKWIADBV-UHFFFAOYSA-N Synonym: benzylamine,benzenemethanamine,monobenzylamine,alpha-aminotoluene,phenylmethyl amine,aminomethyl benzene,1-phenylmethanamine,moringine,n-benzylamine,phenylmethylamine PubChem CID: 7504 ChEBI: CHEBI:40538 IUPAC Name: phenylmethanamine SMILES: C1=CC=C(C=C1)CN
Benzylamine 99.0+%, TCI America™
CAS: 100-46-9 Molecular Formula: C7H9N Molecular Weight (g/mol): 107.156 MDL Number: MFCD00008106 InChI Key: WGQKYBSKWIADBV-UHFFFAOYSA-N Synonym: benzylamine,benzenemethanamine,monobenzylamine,alpha-aminotoluene,phenylmethyl amine,aminomethyl benzene,1-phenylmethanamine,moringine,n-benzylamine,phenylmethylamine PubChem CID: 7504 ChEBI: CHEBI:40538 IUPAC Name: phenylmethanamine SMILES: C1=CC=C(C=C1)CN
Benzylamine, 98+%
CAS: 100-46-9 Molecular Formula: C7H9N Molecular Weight (g/mol): 107.156 MDL Number: MFCD00008106 InChI Key: WGQKYBSKWIADBV-UHFFFAOYSA-N Synonym: benzylamine,benzenemethanamine,monobenzylamine,alpha-aminotoluene,phenylmethyl amine,aminomethyl benzene,1-phenylmethanamine,moringine,n-benzylamine,phenylmethylamine PubChem CID: 7504 ChEBI: CHEBI:40538 IUPAC Name: phenylmethanamine SMILES: C1=CC=C(C=C1)CN
LiChropur™ Benzylamine, For GC Derivatization, 99.0%, MilliporeSigma™ Supelco™
CAS: 100-46-9 Molecular Formula: C7H9N Molecular Weight (g/mol): 107.16 MDL Number: MFCD00008106 InChI Key: WGQKYBSKWIADBV-UHFFFAOYSA-N Synonym: alpha-Aminotoluene IUPAC Name: 1-phenylmethanamine SMILES: NCC1=CC=CC=C1
Sigma Aldrich Benzylamine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Benzylamine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
MP Biomedicals, Inc Benzylamine, MP Biomedicals
CAS: 100-46-9 Molecular Formula: C7H9N Molecular Weight (g/mol): 107.156 InChI Key: WGQKYBSKWIADBV-UHFFFAOYSA-N Synonym: benzylamine,benzenemethanamine,monobenzylamine,alpha-aminotoluene,phenylmethyl amine,aminomethyl benzene,1-phenylmethanamine,moringine,n-benzylamine,phenylmethylamine PubChem CID: 7504 ChEBI: CHEBI:40538 IUPAC Name: phenylmethanamine SMILES: C1=CC=C(C=C1)CN