Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzylamine, MP Biomedicals

Catalog No. ICN214283 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN214283 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN214283 Supplier MP Biomedicals, Inc Supplier No. 0521428380

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 100-46-9
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
InChI Key WGQKYBSKWIADBV-UHFFFAOYSA-N
Synonym benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine
PubChem CID 7504
ChEBI CHEBI:40538
IUPAC Name phenylmethanamine
SMILES C1=CC=C(C=C1)CN

Specifications

Quantity 100 g
Formula Weight 107.16
Chemical Name or Material Benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.