Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Benzylboronic acid pinacol ester, 96%
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Benzylboronic acid pinacol ester finds application in Pd (0)-mediated [11 C] carbonylation reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzylboronic acid pinacol ester finds application in Pd (0)-mediated [11 C] carbonylation reaction.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Benzylboronic acid pinacol ester finds application in Pd (0)-mediated [11 C] carbonylation reaction.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Specifications
Specifications
Density | 0.98 |
Boiling Point | 65°C (0.15 mmHg) |
Quantity | 25 g |
Solubility Information | Insoluble in water. |
Formula Weight | 218.1 |
Percent Purity | 96% |
Chemical Name or Material | Benzylboronic acid pinacol ester |
Safety and Handling
TSCA | No |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction