Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzylboronic acid pinacol ester, 96%

Catalog No. aah3252303
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 87100-28-5 | C13H19BO2 | 218.103 g/mol

Benzylboronic acid pinacol ester finds application in Pd (0)-mediated [11 C] carbonylation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzylboronic acid pinacol ester finds application in Pd (0)-mediated [11 C] carbonylation reaction.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87100-28-5
Molecular Formula C13H19BO2
Molecular Weight (g/mol) 218.103
MDL Number MFCD05663841
InChI Key YCNQPAVKQPLZRS-UHFFFAOYSA-N
Synonym benzylboronic acid pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-phenylmethyl, acmc-209qek, benzylboronic pinacol ester, benzyl boronic acid pinacol ester, benzylboronic acid, pinacol ester, 2-benzyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 4,4,5,5-tetramethyl-2-phenylmethyl-1,3,2-dioxaborolane
PubChem CID 3864964
IUPAC Name 2-benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CC2=CC=CC=C2

Specifications

Density 0.98
Boiling Point 65°C (0.15 mmHg)
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 218.1
Percent Purity 96%
Chemical Name or Material Benzylboronic acid pinacol ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.