Learn More
Benzyldimethylhexadecylammonium chloride hydrate, 95%, Thermo Scientific Chemicals
Description
Benzyldimethylhexadecylammonium chloride has been used in a study to assess the removal of cationic surfactants from aqueous solutions using activated carbon cloth (ACC). It has also been used in a study to investigate the prototropic behavior of the dye, 7-hydroxy-4-methylcoumarin (7H4MC).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 54°C to 58°C |
Color | White |
Linear Formula | CH3(CH2)15N(CH2C6H5)(CH3)2Cl·xH2O |
Quantity | 250 g |
UN Number | UN3261 |
Beilstein | 3802341 |
Sensitivity | Hygroscopic |
Merck Index | 14,2017 |
Solubility Information | Soluble in water (8.5g/L),ethanol,acetone,ethyl acetate,propylene glycol,sorbitol solutions,ether,glycerol,and carbon tetrachloride. |
Formula Weight | 396.1 |
Show More |
Safety and Handling
Hazard Category | H302-H314-H335 |
Hazard Statement | GHS H Statement H301-H311-H314-H318 Toxic if swallowed. Toxic in contact with skin. Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
EINECSNumber | 204-526-3 |
RTECSNumber | BO6822450 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.