Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyldiphenylphosphine, 99%

Catalog No. p-7050677
Click to view available options
Quantity:
0.5 g
2 g
10 g

CAS: 7650-91-1 | C19H17P | 276.32 g/mol

Employed as a catalyst for Suzuki cross-coupling and antiarthritic agent (inhibitor of cathepsin B). It is also used as catalyst in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7650-91-1
Molecular Formula C19H17P
Molecular Weight (g/mol) 276.32
MDL Number MFCD00014083
InChI Key UZCPNEBHTFYJNY-UHFFFAOYSA-N
Synonym benzyldiphenylphosphine, phosphine, diphenyl phenylmethyl, phosphine, benzyldiphenyl, diphenylbenzylphosphine, benzyl diphenyl phosphane, benzyl-diphenylphosphine, benzyl-diphenyl-phosphane, di phenyl-phenylmethyl phosphane, diphenylphosphionomethyl polystyrene, benzyldiphenylphosphine, polymer-bound
PubChem CID 603920
SMILES C(P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 74°C to 75°C
Quantity 0.5 g
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
IUPAC Name benzyldiphenylphosphane
Formula Weight 276.32
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Benzyldiphenylphosphine
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.