Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzylidenemalononitrile, 98%, Thermo Scientific™

Catalog No. AC164291000
Click to view available options
:
100g; Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2700-22-3
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C10H6N2
Linear Formula C6H5CH=C(CN)2
Quantity 100g
MDL Number MFCD00001855
Beilstein 09, 895
Synonym benzylidenemalononitrile, 2-benzylidenemalononitrile, benzalmalononitrile, benzylidenemalonodinitrile, malononitrile, benzylidene, benzal-malonitril, benzylidene malononitrile, 2,2-dicyano-1-phenylethylene, 2-phenyl-1,1-dicyanoethylene, beta,beta-dicyanostyrene
InChI Key WAVNYPVYNSIHNC-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C=C(C#N)C#N
IUPAC Name 2-benzylidenepropanedinitrile
Molecular Weight (g/mol) 154.172
PubChem CID 17608
Formula Weight 154.17
Percent Purity ≥97.5%
Chemical Name or Material Benzylidenemalononitrile, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 220-283-6
RTECSNumber OO3500000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.