Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyloxyacetaldehyde, 95%, stabilized

Catalog No. AC376070050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC376070050 5 g
AC376070010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC376070050 Supplier Thermo Scientific Chemicals Supplier No. 376070050
Only null left
Add to Cart

CAS: 60656-87-3 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60656-87-3
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00191779
InChI Key NFNOAHXEQXMCGT-UHFFFAOYSA-N
Synonym 2-benzyloxy acetaldehyde, benzyloxyacetaldehyde, benzyloxy acetaldehyde, phenylmethoxy acetaldehyde, acetaldehyde, phenylmethoxy, acetaldehyde,2-phenylmethoxy, acmc-20ahpl, benzyloxy-acetaldehyde, 2-phenylmethoxyethanal
PubChem CID 108989
SMILES O=CCOCC1=CC=CC=C1

Specifications

Density 1.0690g/mL
Boiling Point 118°C to 120°C (13.0 mmHg)
Flash Point 113°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Linear Formula C6H5CH2OCH2CHO
Quantity 5 g
Beilstein 06,III,1476
Specific Gravity 1.069
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in chloroform
IUPAC Name 2-(benzyloxy)acetaldehyde
Formula Weight 150.18
Percent Purity 95%
Chemical Name or Material Benzyloxyacetaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 262-349-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.