Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyloxyacetyl chloride, 95%

Catalog No. p-4228988
Click to view available options
Quantity:
5 g

CAS: 19810-31-2 | C9H9ClO2 | 184.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19810-31-2
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.62
InChI Key QISAUDWTBBNJIR-UHFFFAOYSA-N
Synonym benzyloxyacetyl chloride, 2-benzyloxy acetyl chloride, benzyloxy acetyl chloride, benzyloxyacetylchloride, benzyloxy-acetyl chloride, benzyloxyacetic acid chloride, acetyl chloride, phenylmethoxy, acetyl chloride, benzyloxy, alpha-benzyloxy acetyl chloride, bnoch2cocl
PubChem CID 177085
IUPAC Name 2-phenylmethoxyacetyl chloride
SMILES C1=CC=C(C=C1)COCC(=O)Cl

Specifications

Boiling Point 105°C to 107°C (5.0 Torr)
Color White
CAS Min % 94.0
CAS Max % 100.0
Physical Form Crystals or Powder
Quantity 5 g
Assay Percent Range 94% min. as methyl ester (GC)
Flash Point >110°C
Formula Weight 184.62
Percent Purity 95%
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 1.189
Density 1.1890g/mL
Chemical Name or Material Benzyloxyacetyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.