Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyltriethylammonium bromide, 98+%

Catalog No. AAA1429122
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1429122 100 g
AAA1429114 25 g
AAA1429136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1429122 Supplier Thermo Scientific Chemicals Supplier No. A1429122
Add to Cart

CAS: 5197-95-5 | C13H22BrN | 272.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5197-95-5
Molecular Formula C13H22BrN
Molecular Weight (g/mol) 272.23
MDL Number MFCD00011822
InChI Key CHQVQXZFZHACQQ-UHFFFAOYSA-M
Synonym benzyltriethylammonium bromide, n-benzyl-n,n-diethylethanaminium bromide, benzyl triethyl ammonium bromide, triethylbenzylammonium bromide, bteab, benzenemethanaminium, n,n,n-triethyl-, bromide, benzyltriethyl ammonium bromide, n-benzyl-n,n,n-triethylammoniumbromide, benzyltriethylazanium bromide, pubchem7797
PubChem CID 165294
IUPAC Name benzyl(triethyl)azanium;bromide
SMILES [Br-].CC[N+](CC)(CC)CC1=CC=CC=C1

Specifications

Melting Point ∼200°C (decomposition)
Quantity 100 g
Beilstein 3739113
Sensitivity Hygroscopic
Formula Weight 272.24
Percent Purity ≥98%
Chemical Name or Material Benzyltriethylammonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-986-1
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.