Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyltriethylammonium Iodide 98.0+%, TCI America™
SDP

Catalog No. B044625G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 5400-94-2
Molecular Formula C13H22IN
Molecular Weight (g/mol) 319.23
MDL Number MFCD00043178
InChI Key JWLJBISFJGEYMT-UHFFFAOYSA-M
PubChem CID 122756
IUPAC Name benzyltriethylazanium iodide
SMILES [I-].CC[N+](CC)(CC)CC1=CC=CC=C1

Specifications

Melting Point 171°C
Color White
Quantity 25 g
Formula Weight 319.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyltriethylammonium Iodide
EINECSNumber (3)-2694
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.