Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyltrimethylammonium Dichloroiodate 97.0+%, TCI America™
SDP

Catalog No. B160425G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 114971-52-7
Molecular Formula C10H16Cl2IN
Molecular Weight (g/mol) 348.049
MDL Number MFCD00075259
InChI Key PPDJNZTUDFPAHX-UHFFFAOYSA-N
Synonym benzyltrimethylammonium dichloroiodate, benzyltrimethylammonium dichloroiodide, btma-icl2, benzyl trimethylammonium dichloroiodate, benzyltrimethylazanium; dichloroiodanuide, pubchem21920, benzyltrimethylammoniumdichloroiodate
PubChem CID 2724680
SMILES C[N+](C)(C)CC1=CC=CC=C1.Cl[I-]Cl

Specifications

Melting Point 125°C
Quantity 25 g
Formula Weight 348.05
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyltrimethylammonium Dichloroiodate
EINECSNumber (3)-2694
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.